Direkt Rot 80 Färbemittel für Histologie

Lieferant: RAL

Synonyme: CI 35780, Sirius Red F3B, Sirius Red, Picro-Sirius Red

363440-0005
720-1194EA 0 EUR
720-1194
Direkt Rot 80 Färbemittel für Histologie
Direkt Rot 80

Sirius red is a laboratory reagent.


  • Dark red brown crystalline powder
  • Life time 120 months


Designed for applications in histocytology.

Formel: C₄₅H₂₆N₁₀Na₆O₂₁S₆
Molecular Weight: 1373,051 g/mol
MDL: MFCD00054389
CAS-Nummer: 2610-10-8
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR