Urine test strips, Albustix®
Lieferant: BAYER VITAL
2872 2614217
720-1339EA 40.2 EUR
720-1339 BAYR2614217
Urine test strips, Albustix®
Klinisches Diagnosezubehör Klinische Reagenzien und Kits
Reagent strips for urinalysis tests.
  • For checking protein levels in urine
  • Help to provide information on kidney function
  • For in vitro diagnostic use