Urine test strips, Albustix®
Lieferant: BAYER VITAL
2872
2614217
720-1339EA
40.2
EUR
720-1339
BAYR2614217
Urine test strips, Albustix®
Klinisches Diagnosezubehör
Klinische Reagenzien und Kits
Reagent strips for urinalysis tests.
- For checking protein levels in urine
- Help to provide information on kidney function
- For in vitro diagnostic use