Inbedmedium HiQA Pertex™

Lieferant: CELLPATH

SEA-0100-00A
720-2343EA 173 EUR
720-2343
Inbedmedium HiQA Pertex™
Färbemittel Mikroskopie- und Färbereagenzien

Pertex mounting medium is neutral, colourless and made of a resin, based on butyl acrylate polymer dissolved in xylene. This provides a high embedding quality and enables fast drying.


  • Low viscosity formulation, ideal for automatic cover slipping instruments
  • Slides exhibit minimal discolouration or drying back
  • Refractive index: 1.992 at 20 °C if damp
  • Storage life: 2 years


Supplied in a wide-necked, tamper-evident PET bottle.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR