Inbedmatrix OCT - Cryo
Lieferant: CELLPATH
KMA-0100-00A
720-2356EA 24.1 EUR
720-2356
Inbedmatrix OCT - Cryo
Färbemittel Mikroskopie- und Färbereagenzien
‘OCT’ cryo microtomy embedding medium is designed for use at –20 ˚C to –30 ˚C. The medium freezes rapidly with minimal damage by crystal formation and is an ideal support medium for cutting tissue sections.
  • Liquid level clearly visible in bottle
  • Excellent quality, highly viscous
  • Easy to use dispenser bottle
‘OCT’ is easily removed from the microscope slide prior to staining. The 125 ml bottle is a convenient size with a nozzle moulded into the cap.