Blood collection tubes with EDTA anticoagulant, vacuum, Vacutest®
Lieferant: KIMA SAS DI CHIARIN RENZO
13030 13125 135400 13560 13040 13001 13005 13505 13060 135100 135300 13010 13510
720-2742EA 257 EUR
720-2742 KIMA13125 720-2745 720-2770 720-2743 720-2765 720-2766 720-2768 720-2767 KIMA135100 KIMA135300 KIMA13010 KIMA13510
Blood collection tubes with EDTA anticoagulant, vacuum, Vacutest®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in different sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
More Product Information