Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: HFPB, Tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]boron sodium

M-Clarity™ Qualitätsstufe = MQ100

72015-50MG
72015-50MGEA 246 EUR
72015-50MG
Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate, Selectophore™ Ionophor, Supelco®
Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate

Lipophilic ion exchanger for ion-selective membrane electrodes.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₅₆H₃₆BF₄₈NaO₈·3H₂O
Molecular Weight: 1836,65 g/mol
MDL: MFCD01863091
CAS-Nummer: 120945-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5557a980c27b81b29dd06c19672eeda981f1eac0&vid=67119c7b699a459e98a07539f81f823c8ba85927&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR