Neocuproin Hemihydrat, Supelco®

Lieferant: Merck

Synonyme: 2,9-Dimethyl-1,10-phenanthrolin Hemihydrat

M-Clarity™ Qualitätsstufe = MQ100

72080-1G
72080-1GEA 28.1 EUR
72080-1G
Neocuproin Hemihydrat, Supelco®
Neocuproin Hemihydrat
Formel: C₂₈H₂₆N₄O
Molecular Weight: 434,53 g/mol
MDL: MFCD00149306
CAS-Nummer: 34302-69-7
EINECS: 207-601-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b51050a32d317202b2f595c13f8e780d2d195c96&vid=b2f6b5d8f308df518603e585ca990ea7ebe8890f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR