Neocuproin Hydrochlorid Monohydrat, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
72090-1G
72090-5G
72090-1GEA
45.8
EUR
72090-1G
72090-5G
Neocuproin Hydrochlorid Monohydrat, Supelco®
Formel:
C₁₄H₁₃ClN₂·1H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012645 CAS-Nummer: 303136-82-5 EINECS: 230-732-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...