Nitrite ion standard solution, for ion-selective electrodes, Supelco®
Lieferant: Merck
72586-100ML
72586-100MLEA 51 EUR
72586-100ML
Nitrite ion standard solution, for ion-selective electrodes, Supelco®
Standards Anionische Standards Nitrit-Standards
Prepared with NaNO₂ and H₂O.
Bitte beachten Storage Temperature: 2 to 8 °C
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=571bda99f678f042329b37c875054298e2c2451f&vid=571bda99f678f042329b37c875054298e2c2451f&prid=web&clid=SAPDAM