Nitrite ionophore I, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyme: Cyanoaqua-cobyrinic acid heptakis(2-phenylethyl ester)

M-Clarity™ Qualitätsstufe = MQ100

72590-10MG
72590-10MGEA 790 EUR
72590-10MG
Nitrite ionophore I, Selectophore™, Funktion getestet Ionophor, Supelco®
Nitrite ionophore I

Ionophore for nitrite-selective liquid membrane electrodes.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₀₂H₁₁₇ClCoN₅O₁₉
Molecular Weight: 1811,43 g/mol
MDL: MFCD00213827
CAS-Nummer: 130549-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5005a178d5bf1fc10eb339ec301af30fe55612bd&vid=e2426bfc52ba4d42e05e3e3c20b53378a70dc248&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR