Arsenic standards for ICP, TraceCERT®, Supelco®
Lieferant: Merck
76686-100ML
72718-100ML
76686-100MLEA
111
EUR
76686-100ML
72718-100ML
Arsenic standards for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Arsen-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.