Multiple horizontal gel electrophoresis system, Owl™ A2-OK
Lieferant: THERMO OWL SCIENTIFIC
A2-OK
730-0161EA 3590 EUR
730-0161
Multiple horizontal gel electrophoresis system, Owl™ A2-OK
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
Owl™ A2-OK multiple gel system is ideal for teaching labs or labs that need to run multiple gels simultaneously.
  • Up to six mini gels can be run
  • One teacher can instruct up to 12 students with one system and only one power supply
  • The system can screen up to 144 samples at once when used with optional combs (120 with supplied combs)
  • EasyCast casting design allows the casting of gels without tape or plugs
  • UV transmissible gel trays may be placed directly on the transilluminator for photodocumentaion
Lieferumfang: Each unit includes buffer chamber with 3-point levelling base, SuperSafe lid with attached power supply leads, EasyCast gasketed UVT gel trays (6), external casting chambers (2), gel tray holder, and 12 double-sided combs (10-well, 1,0/1,5 mm thick).