Deep well plates, 96-well, Bioblock™
Lieferant: Simport Scientific
T110-6
730-1091EA 286 EUR
730-1091
Deep well plates, 96-well, Bioblock™
Mikrotiterplatten Deep-Well-Mikroplatten
PS. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) can withstand centrifugation up to 3000×g. It is not DMSO resistant.
Lieferumfang: Packed in bags of 4 plates.