X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Lieferant: VWR Collection
730-1498EA
0
EUR
730-1498
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Substrat für den Nachweis von β-Galactosidase.
Formel:
C₁₄H₁₅BrClNO₆ |
MDL:
MFCD00005666 CAS-Nummer: 7240-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...