Supelco® TraceCERT® Organic Reference Standard, 2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
Lieferant: Merck
73096-50MG
73096-50MGEA
82
EUR
73096-50MG
Supelco® TraceCERT® Organic Reference Standard, 2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
Standards
Organische Standards
- GHS signal: GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338