Electrofusion buffer, BTXpress Cytofusion® Medium C
Lieferant: BTX
47-0001
731-0348EA
572
EUR
731-0348
Electrofusion buffer, BTXpress Cytofusion® Medium C
Nukleinsäure-Reagenzien
Transformations-Reagenzien
BTXpress Cytofusion® Medium C is an advanced electrofusion buffer designed especially for hybridoma production. The low conductivity buffer is specially formulated to minimise cell turbulence during cell alignment and heating during electrofusion, for robust cell fusion efficiency and high cell viability. BTXpress Cytofusion® Medium C is sterile filtered from the highest quality non animal, medical grade reagents.