Transfection reagent, TransIT®-Insect
Lieferant: MIRUS
TransIT®-Insect transfection reagent is an animal-origin-free transfection reagent specifically optimised for high gene expression in a variety of insect cell types.
- Exceptional DNA delivery in insect cell types, including Sf9, High Five™ and S2
- Can also be used to transfect insect cells with baculovirus DNA to make recombinant virus
TransIT®-Insect Transfection Reagent is in 80% ethanol and should be stored tightly capped at –20 ºC.
TransIT-Insect can also be used to transfect insect cells with baculovirus DNA to make recombinant virus. TransIT-Insect is composed of animal-origin free components and is serum compatible, which eliminates the need for any culture medium change after transfection.
Bestellinformation: TransIT® -Insect Transfection Reagent is available in 0,4 ml, 1 ml, 5×1 ml (5-pack) and 10×1 ml (10-pack) configurations.