Petri dish electrode

Lieferant: BTX
45-0100
732-0000EA 3460 EUR
732-0000
Petri dish electrode
Elektroden Elektroporationselektroden
This petri dish electrode is designed to be used with a 100 mm petri dish that functions as the electroporation chamber. The electrode is used to electroporate adherent cells or tissue grown in a petri dish.

  • Easy to use: Users simply need to add the exogenous molecule of interest into the electroporation buffer
  • Electrode assembly contains parallel stainless steel electrodes which generate a homogeneous field
  • Petri dish electrode is compatible with most BTX Generators

Typical applications include adherent mammalian cell transfections and plant tissue cell transfections.

Generator compatibility: ECM 830, ECM 630 and ECM 2001
Voltage range: 0 to 2000 V

Pulse length range: 10 µsec to 10 msec

Volume range: 10 to 50 ml

Electrode gap: 2 mm
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR