Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Lieferant: EPPENDORF
0030128.508 0030128.516 0030128.524 0030128.532 0030128.540 0030128.940 0030128.974 0030128.958 0030128.931 0030128.966 0030129.342 0030129.350 0030129.628 0030113.578
732-0101EA 224 EUR
732-0101 732-0102 732-0103 732-0104 732-0105 EPPE0030128.940 EPPE0030128.974 EPPE0030128.958 EPPE0030128.931 EPPE0030128.966 737-0183 737-0184 EPPE0030129.628 737-0257
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Mikrotiterplatten PCR-Platten
Eppendorf's twin.tec® 384-well, 45 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimising the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
The Eppendorf twin.tec® PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Twin.tec plates are available in many different formats to suit different workflows and applications, for example: skirted, semi-skirted, and unskirted; divisible plates to avoid waste in small assays; individually wrapped forensic DNA grade plates; microbiology plates; barcoded plates.
All Eppendorf twin.tec PCR plates are 'PCR clean', having been batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors. With twin.tec PCR plates you'll have maximum confidence in your results.
Barcoded option provides 3-level coding for safe sample and vessel identification. ECC 200 error correction function enables safe code reading with up to 30 % damaged codes.
Autoclavable: 121 °C, 20 min.
Centrifugation stability: 2250×g