The DigiDoc-It® 130 imaging system connects to a computer (not included) and is designed as a simple and economical system for acquisition and documentation of gels and plates. The system combines a digital colour camera with megapixel resolution housed in the top of a compact, lightweight hood.
- Gels can be viewed through the UV blocking viewpoint, which for maximum darkness can be closed when taking pictures
- Exclusive capture software controls camera functions, image capture and image enhancement, annotation and archiving, and enables live preview of images
- Software allows selection of user defined default settings and templates
- Lightweight door opens for access to the UV transilluminator surface (transilluminator not supplied)
- Handles allow hood to be easily removed from the transilluminator
Haube (B×T×H): 451×305×422 mm
Lieferumfang: Each system includes megapixel digital camera (DigiCam 130), hood enclosure, control software and ethidium bromide filter. An external PC is required but not supplied.