Leere Nachfüll-Racks
Lieferant: Thermo Fisher Scientific
2004-ER 2006-ER 2008-ER
732-1171EA 20.5 EUR
732-1171 732-1170 732-1169
Leere Nachfüll-Racks
Racks und Boxen für Pipettenspitzen
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ pipette tip empty hinged racks.
  • Certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Front latch closes and locks, ensuring safe and secure storage throughout