Pipette tips, gel loading, Axygen®
Lieferant: Corning
TGL-10FT-17-R TGL-200FT-37-R TGL-200FT-37RS TGL-200RD-57-R TGL-200RD-57RS
732-1408EA 186 EUR
732-1408 732-1412 732-1413 732-1414 732-1415
Pipette tips, gel loading, Axygen®
Pipettenspitzen
These gel loading tips made of transparent PP are designed to fit a wide variety of single and multi channel pipettors, compatible with most popular brand of pipettors. Available in either flat or round versions, with different outer diameters for different gel thickness.
  • Universal fit for research grade pipettors
  • Non pyrogenic and certified to be RNase-/DNase-free
  • Tips with 0,17 mm Ext.Ø. are for loading 0,2 mm thick gels
  • Tips with 0,37 mm Ext.Ø. are for loading 0,4 mm thick gels
  • Pipette tips with 0,57 mm Ext.Ø. have 2 and 5 µl graduations for quick volume check