PCR tubes, 0,5 ml, without cap, Axygen®
Lieferant: Corning
TUB-310-NC
732-1416EA 117.46 EUR
732-1416
PCR tubes, 0,5 ml, without cap, Axygen®
Röhrchen PCR-Gefäße
PCR tube, made of PP, designed to fit standart 96 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer and even insertion cap ensures a tight fit to prevent evaporation.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatible with ABI-310 Instrument