These multi-barrier filter pipette tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact assuring the valuable samples and research have not been compromised.
Effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material
Proprietary filter design prevents possible PCR inhibition
Maxymum Recovery® technology for low sample retention and high yield