Barrier tips, GEN3™, Axygen®
Lieferant: Corning
GEN3-1000-L-RS
732-7515EA 704 EUR
732-7515
Barrier tips, GEN3™, Axygen®
Pipettenspitzen
These multi-barrier filter pipette tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact assuring the valuable samples and research have not been compromised.
  • Effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material
  • Proprietary filter design prevents possible PCR inhibition
  • Maxymum Recovery® technology for low sample retention and high yield
  • Hinged rack for convenience and ease of use
  • Certified DNase-/RNase-free and non pyrogenic
  • Certified human gDNA and PCR inhibition free