VWR® Probe One-Step RT-qPCR Master Mix with UDG
Lieferant: VWR Chemicals
73224-0500EA 819 EUR
73224-0500 73224-0100 73224-1000 73224-2500
VWR® Probe One-Step RT-qPCR Master Mix with UDG
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
VWR® Probe One-Step RT-qPCR Master Mix with UDG is designed for the detection and quantitation of RNA targets and is an ideal choice for multiplex applications. It features a distinctive universal passive reference dye that is compatible with ROX-dependent and ROX-independent qPCR instruments.
  • For the detection and quantification of RNA targets using probe-based technology, such as TaqMan®
  • Ready-to-use master mix containing all necessary reaction components, except for primers, probes, and RNA template
  • For multiplex applications of up to 5 targets
  • Suitable for a variety of RNA sample types
  • Includes Hot Start Taq DNA Polymerase and VWR® Reverse Transcriptase for improved reaction specificity and room temperature setup
  • Includes dUTP and UDG to prevent carryover contamination
  • Supplied at a 4X concentration to accomodate higher sample volume inputs
The VWR® Probe One-Step RT-qPCR Master Mix with UDG is offered in a convenient single-tube format, and includes the temperature-controlled VWR® Reverse Transcriptase and Murine RNase Inhibitor to assist in preventing RNA degradation. This temperature-controlled RT enzyme prevents unwanted non-specific activity and displays superior thermostability in comparison to other reverse transcriptases.
Bitte beachten Store at −20 °C. For research use only.