Wasser, AccuGENE® Molecular_Biology
Lieferant: LONZA
Synonyme:
Wasserstoffoxid, Hydrogenoxid
AccuGENE™ Molecular Biology Water is ideal for a wide range of molecular biology applications. 18 megOhm water is filtered using a 0.2 micron filter, and filled into sterile bottles.
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
- High Quality - No detectable DNase, RNase, or protease activities.
- Efficient - Ready-made solutions eliminate experiment preparation time.
- Flexible - Customized solutions are available to meet individual needs.
Water is a key component in so many experiments and pure, quality controlled and traceable water can help you improve your control over critical, sensitive experiments. Lonza’s AccugeneTM Molecular Biology Grade water has been the reagent of choice for decades for electrophoresis, cloning, PCR and other applications that demand the strictest of quality controls.
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 Schmelzpunkt: 0 Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...