Hybridisation buffer, SSPE 20X, AccuGENE™
Lieferant: LONZA
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
- Ready to use solutions
- Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
- Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.