Hybridisation buffer, SSPE 20X, AccuGENE™
Lieferant: LONZA
51214
733-1638EA 58.8 EUR
733-1638
Hybridisation buffer, SSPE 20X, AccuGENE™
Puffer Elektrophorese-Puffer Hybridisierungspuffer
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Ready to use solutions
  • Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
  • Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.