Wasser, AccuGENE® Molecular_Biology

Lieferant: LONZA

Synonyme: Wasserstoffoxid, Hydrogenoxid

51200 51224
733-1631EA 37.1 EUR
733-1631 733-1643
Wasser, AccuGENE® Molecular_Biology
Wasser

AccuGENE™ Molecular Biology Water is ideal for a wide range of molecular biology applications. 18 megOhm water is filtered using a 0.2 micron filter, and filled into sterile bottles.


  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs.


Water is a key component in so many experiments and pure, quality controlled and traceable water can help you improve your control over critical, sensitive experiments. Lonza’s AccugeneTM Molecular Biology Grade water has been the reagent of choice for decades for electrophoresis, cloning, PCR and other applications that demand the strictest of quality controls.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR