White light box
Lieferant: EDVOTEK
552
733-2003EA 252 EUR
733-2003
White light box
Beleuchtung Leuchtkästen
The Edvotek White Light Box features a spacious 25×25 cm viewing area illuminated by long life LEDs and is housed in a slim aluminium body. It is designed to safely enhance the visualisation of DNA stained with FlashBlue™, proteins stained with Coomassie Blue and autoradiograms.
Power: 8 W
Dual Voltage: 110/220 V, 50/60 Hz
Dimensions: 345×248 mm