PerfeCTa® qPCR FastMix® II
Lieferant: Quantabio
95118-012 95118-05K 95118-250 95120-012 95120-05K 95120-250 95119-012 95119-05K 95119-250
733-2108EA 1360 EUR
733-2108 733-2109 733-2110 733-2114 733-2115 733-2116 733-2111 733-2112 733-2113
PerfeCTa® qPCR FastMix® II
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
PerfeCTa® qPCR FastMix® II is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments. It is a versatile and robust solution that provides the ultimate sensitivity and high PCR efficiency using a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes. PerfeCTa® qPCR FastMix® II is provided as a 2X concentrated ready to use reaction cocktail that contains all required reaction components, except primers, probe(s), and DNA template. The light blue colour of the AccuVue™ tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimise pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.
A key component of PerfeCTa® qPCR FastMix® II is an ultra-pure, processive thermostable DNA polymerase that is free of detectable E. coli DNA. PerfeCTa® qPCR FastMix® II is ideal for demanding qPCR applications such as bacterial pathogen detection where residual host DNA in typical recombinant enzyme preparations can limit assay sensitivity and obscure detection of low copy samples. The enzyme in PerfeCTa® qPCR FastMix® II is combined with high avidity monoclonal antibodies to provide a stringent automatic Hot Start that allows reaction assembly, and temporary storage, at room temperature prior to PCR amplification.
Lieferumfang: Kits are available with or without ROX reference dye. All kits comprise of 2X reaction buffer containing optimised concentrations of MgCl₂, dNTPs (dATP, dCTP, dGTP, dTTP), Hot Start DNA polymerase, AccuVue™ blue qPCR dye, and stabilisers.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Anwendung qPCR Probe