VWR®, DNA ladders
Lieferant: VWR Chemicals
733-2602EA
50.8
EUR
733-2602
733-2601
733-2603
VWR®, DNA ladders
Elektrophorese-Reagenzien
Molekulargewichtsmarker
VWR® DNA ladders are convenient dsDNA ladders supplied in a loading buffer, which are ready to use on agarose and SDS DNA gels.
- Ready to use loading buffer
- For direct loading and easy visualisation
- Suitable with TBE and TAE electrophoresis systems
- Clear, distinct bands
- Blue dye front running at 100 to 300 bp at 0,5 to 1,5% agarose
Bestellinformation: Supplied in 0,5 ml packs for 100 lanes.