VWR®, Taq DNA polymerase, glycerol-free
Lieferant: VWR Chemicals
Highly concentrated Taq DNA polymerase glycerol free 50 U/µl is ideal for industrial use and manufactures of diagnostic PCR kits.
- Suitable for automated high throughput testing
- Suitable for applications involving freeze drying
- Suitable for robot-aided pipetting
- Ideal for kit manufacturers
- Concentration: 50 U/µl
- Fidelity: 1×Taq
- Amplicon size: <5 kb
Taq DNA Polymerase Glycerol Free 50 U is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.
Taq DNA Polymerase Glycerol Free has both a 5'→3' DNA polymerase and a 5'→3' exonuclease activity. The enzyme lacks a 3'→5' exonuclease activity (no proofreading ability). Taq DNA Polymerase Glycerol Free leaves an A' overhang, which makes the enzyme ideal for TA cloning.
The glycerol free formulation makes it especially well-suited for freeze drying and automatisation.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...