Dibutylammonium acetate 0.5 M in Wasser, LiChropur™ für die Ionenpaar-Chromatographie, Supelco®
Lieferant: Merck
Dibutylammonium acetate is an ion-pairing agent with quaternary ammonium ions. It generally helps in ionization of carbohydrate fragments providing a volatile solvent for the mass spectrometric technique.
Verpackung: Concentrate in ampules for 6x1 l 0.005 M ready-to-use solution
Formel:
C₈H₁₉N |
MDL:
MFCD00191381 CAS-Nummer: 19070-91-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...