Petrischalen, gefärbt, Sterilin®
Lieferant: Sterilin
101VBLUE 101VRED
734-0458EA 330 EUR
734-0458 734-0459
Petrischalen, gefärbt, Sterilin®
Zellkulturschalen Petrischalen
PS, mit Deckel. Farbkodierte Petrischalen besonders zur Kennzeichnung von Gruppenarbeiten in Lehrlaboren geeignet.
  • Aids ease of identification
  • Aseptically manufactured under cleanroom conditions (class 7 ISO14644) to exclude microbiological contamination
  • Produced and tested in accordance with the BS611 part 2 standard
  • Manufactured using cadmium free non cytotoxic colourants
Aseptically manufactured, In vitro use only. Perform non standard applications with Thermo Scientific™ Sterilin™ special Petri dishes. minimum order quantities apply. Not available in America.