Collection and analysis plates, Whatman UNIPLATE™
Lieferant: Whatman products (Cytiva)
7701-5110
734-1217EA 405 EUR
734-1217
Collection and analysis plates, Whatman UNIPLATE™
Mikrotiterplatten Deep-Well-Mikroplatten
PP, with clear PS lid. The Whatman™ UNIPLATE collection and analysis microplates accomodate 24 well plates, have round bottom wells.
  • Chemical compatibility: Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage. Opaque plates prevent optical cross talk in light emitting assays
  • Guaranteed for use with robotic handlers and centrifuge carriers
Standards: UNIPLATE microplates conform to the ANSI/SBS microplate standard and fit most microplate readers and automated plate handling devices.