Optical bottom plates, 96-well, CC²™ coated, Nunc™
Lieferant: Thermo Fisher Scientific
160376
734-1354EA 3200 EUR
734-1354
Optical bottom plates, 96-well, CC²™ coated, Nunc™
Mikrotiterplatten Zellkultur-Platten
PS with coverglass base, sterile, with lid. CC2™ surface treatment closely mimics a biological surface similar to Poly-D-Lysine and is a superior surface for attachment and growth of fastidious cells.
  • White or black upper structure with coverglass bottom for minimum light scatter and low auto-fluorescence, ensuring accurate results due to higher signal-to-noise ratios
  • Optimum clarity for viewing well contents