Corning® Matrigel® hESC-Qualified Matrix, Corning
Lieferant: Corning
Pre-screned for compatibility with mTeSR™1 medium by StemCell Technologies providing the reproducibility and consistency essential for hESC and iPSC feeder-free stem cell culture.
- Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
- Extensive PCR testing is performed to screen for a number of pathogens
- Tested and found negative for bacteria, fungi, and mycoplasma
- Protein concentrations determined by Lowry method
- Endotoxin units measured by Limulus Amoebocyte Lysate assay
- Matrigel Matrix gel stability tested for a period of 14 days at 37°C
- Biological activity determined for each lot using a neurite outgrowth assay.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...