96 MicroWell™ Platten, Nunclon™∆
Lieferant: Thermo Fisher Scientific
143761 156545 161093 163320 167008 168055 168136
734-2033EA 138 EUR
734-2033 732-2601 734-2073 734-2080 734-2097 732-2607 734-1192
96 MicroWell™ Platten, Nunclon™∆
Mikrotiterplatten Zellkultur-Platten
PS, erhöhter Rand, steril. Ideal für Zellkultur, Klonierungen, Virustitration und Zellfusion.
  • Plate edges and lids designed to reduce evaporation
  • Raised well rims reduce risk of cross contamination
  • Lids shaped to facilitate stacking and handling
  • 96-well footprint fits standard equipment
Farbe: Transparent
Externe L×B: 128×86 mm
External L×W: 128×86 mm