Nunc™ EasYDish™ Cell Culture Dishes with Nunclon™ Supra Surface
Lieferant: Thermo Fisher Scientific
150470 150472 150474 150476 150478
734-3641EA 8 EUR
734-3641 734-3638 734-3643 734-3636 734-3639
Nunc™ EasYDish™ Cell Culture Dishes with Nunclon™ Supra Surface
Zellkulturschalen Zell- und Gewebekultur-Platten
Experience optimal performance with Thermo Scientific Nunc™ EasYDish™ dishes with Nunclon™ Supra surface treatment for culturing a wide variety of fastidious adherent cells.
  • The beveled grip ring is easy to hold and stays secure in your gloved hand
  • Available in four sizes: 35, 60, 100, and 150 mm
  • Sterile, not autoclavable
  • Clear, round, flat bottom, not gridded
  • Single use
  • Consistent lot-to-lot performance
  • Non-cytotoxic and non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • 5 years of shelf life
Intended for general cell culture applications, the Nunclon™ Supra surface is more hydrophilic than the Nunclon™ Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon™ Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon™ Supra cultureware is a great xeno-free solution for cell therapy research applications.