Disposable round bottom culture tubes, screw cap style, bulk pack, PYREX®
Lieferant: Corning
99449-13 99449-16 99449-16X 99449-20 99449-20X
734-4239EA 438.02 EUR
734-4239 734-4240 734-4241 734-4243 734-4244
Disposable round bottom culture tubes, screw cap style, bulk pack, PYREX®
Röhrchen Kulturröhrchen
A round bottom tube without marking spot. Tubes are made of borosilicate glass and are sold without caps.