Multidishes, non treated, Nunc™
Lieferant: Thermo Fisher Scientific
179820 144530 150200 150239 150787
734-2375EA 329 EUR
734-2375 734-0994 735-0223 735-0224 735-0225
Multidishes, non treated, Nunc™
Mikrotiterplatten Zellkultur-Platten
PS, non treated, with lid, sterile.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Non pyrogenic