Advanced multimode microplate reader, Spark®
Lieferant: Tecan
93000001
735-0399EA 0 EUR
735-0399
Advanced multimode microplate reader, Spark®
Mikrotiterplatten-Photometer
Advanced multimode microplate reader with Monochromator or Fusion optics for 6 to 1536 well plates.
  • High Speed Monochromators for fast wavelength scanning
  • Adanced Fusion Optics combines the flexibility of monochromators with the sensitivity of filters for all fluorescence measurements
  • Bright field cell imaging option for counting a broad range of cells in suspension using Tecan's Cell Chip™
  • Dedicted luminescence module with photon counting PMT, multicolor filter wheel for luminescence scanning and multicolor luminescece assays
  • Fully integrated Gas Control Module with CO₂ and O₂ regulation for cell kinetic studies
  • Humidity cassette for evaporation protection
  • Integrated lid-lifting system
  • Supports 6 to 1536 well plates in all detection modes with enhanced reading modules
  • Bright field imaging option supports 6 to 384 well SBS plates
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample in parallel
  • A260/280 purity check
  • Labeling effiency for nucleic acids
  • Full-spectra scan (Spark, SparkCyto)
The advanced Spark multimode microplate reader offers solutions for virtually any life science research or drug discovery applications. Spark can be both freely configured and upgraded in the future, providing you an instrument that can meet your needs today and in the future.
Spark's Te-Cool™ module allows to perform assays at the specified temperature in the reader for long periods, even below ambient temperature.
Precise cooling using Te-Cool empowers optimal performance of enzymatic assays, Alpha Technology® assays, or kinetic assays using live organisms.
NanoQuant Plate allows parallel quantification and analysis of up to 16 nucleic acid or protein samples, in sample volumes as little as 2 μl. For convenience and optimal data quality, the Nano-Quant Plate is the only low volume plate on the market that is 100% calibration free.
Standards: Carbon neutral certificate (ClimatePartner).
Bestellinformation: Accessories not included (must be ordered seperately). Additional configuration available on request.
Lieferumfang: Spark is supplied with SparkControl™ software, user manuals as pdf-files, and power cable.
Bitte beachten Spark is for Research Use Only. Not for use in diagnostic procedures.