Crystallisation plates, CrystalStar™, 24-well CombiPlate™ and CrystalBridge™
Lieferant: Greiner Bio-One
662050_24
737-3508EA 479 EUR
737-3508
Crystallisation plates, CrystalStar™, 24-well CombiPlate™ and CrystalBridge™
Mikrotiterplatten Kristallisationsplatten
The CombiPlate™ is a universal 24 well crystallisation plate.
  • Clear PS combined with a flat distortion free bottom offers excellent optical properties
  • A flattened raised ring around each well reduces the risk of cross contamination and makes it possible to seal the wells with silicone grease and coverslips (Ø18 mm) or VIEWseal™ sealer
  • Slightly raised lid protects the coverslips and sealer during transport and storage
  • For sitting drop experiments, CrystalBridge™ inserts fit exactly into the wells of the CombiPlate™