2-Nitrophenyloctylether, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

73732-100ML 73732-25ML 73732-5ML
73732-100MLEA 790 EUR
73732-100ML 73732-25ML 73732-5ML
2-Nitrophenyloctylether, Supelco®
2-Nitrophenyloctylether
Formel: C₁₄H₂₁NO₃
MDL: MFCD00014693
CAS-Nummer: 37682-29-4
EINECS: 253-623-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e6ed89130a68325e3a9cfdf571ad0a357e9a2a8&vid=42fd4c23719ac0a005a462640d6cf743fd7abf9a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR