Filterplatten, 96-Well, für DNA-Aufreinigung, AcroPrep™ Advance
Lieferant: Cytiva (Formerly Pall Lab)
8132 8032
738-0120EA 171 EUR
738-0120 738-0108
Filterplatten, 96-Well, für DNA-Aufreinigung, AcroPrep™ Advance
Mikrotiterplatten Mikrofiltrationsplatten
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification, and general filtration.
  • High binding capacity - optimised for maximum binding and yield of DNA from a variety of sample types
  • Well geometry results in reduced hold-up volume and high recovery of DNA
  • Outlet tip geometry provides direct flow of samples into receiver plate without concerns about cross contamination
  • Optimised membrane ensures high yields of contaminant-free DNA for downstream applications
  • Automation compatible - manufactured in accordance with SBS guidelines, which allows entire DNA purification process to be performed on automated equipment
AcroPrep™ Advance 96-Well-Filterplatten für DNA-Aufreinigung: Anwendungen umfassen Plasmid-DNA-Aufreinigung sowie Aufreinigung genomischer DNA.