Storage plates for liquid acoustic dispensing, 384- and 1536-well, LoBase
Lieferant: Greiner Bio-One
783860-906
738-0180EA 3900 EUR
738-0180
Storage plates for liquid acoustic dispensing, 384- and 1536-well, LoBase
Mikrotiterplatten Deep-Well-Mikroplatten
COC, clear, non sterile, without lid. Made of solvent resistant cycloolefin, these microplates are characterised by good chemical resistance to polar solvents, such as DMSO, and alcohols. They are designed for acoustic liquid handling.
  • Small volumes can be pipetted very precisely and with minimal disturbance without the risk of cross contamination
  • Free from DNase, RNase and human DNA and non pyrogenic
L×W (mm): 127,76×85,48 mm