Filter plates, 96-well, with long tip filter, for nucleic acid binding (NAB), AcroPrep™ Advance
Lieferant: Cytiva (Formerly Pall Lab)
8133
738-0209EA 169 EUR
738-0209
Filter plates, 96-well, with long tip filter, for nucleic acid binding (NAB), AcroPrep™ Advance
Mikrotiterplatten Mikrofiltrationsplatten
The Pall AcroPrep™ Advance 96-well long tip filter plate for nucleic acid binding (NAB) incorporates a silica-based quartz glass fibre media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples. This media offers researchers the flexibility to purify plasmid DNA from bacteria, and genomic DNA or total RNA from cell culture samples using a single plate.
  • Single plate for multiple applications reduces chance of cross contamination - critical for reproducible, high quality results
  • Single plate aids smooth sample processing flow
  • Long tip design minimises hanging drop formation