BIOFLOAT™ 384-Well Cell Culture Spheroid Plate (U-bottom)
Lieferant: FACELLITATE
F224384-2
738-0261EA 460 EUR
738-0261
BIOFLOAT™ 384-Well Cell Culture Spheroid Plate (U-bottom)
Mikrotiterplatten Zellkultur-Platten
The BIOFLOAT™ 384-well plate outperforms existing products by its anti-adhesive properties.
  • Rapid: Spheroids are formed very fast
  • Round: The spheroids are highly uniform and allow for perfect data consistency
  • Reliable: The reliable quality of the BIOFLOAT™ surface makes it a premium product for spheroid and organoid culture
The BIOFLOAT™ 384-well plate provides a highly defined, fully inert, and cell-repelling surface.
Temperature range: – 20 to + 50 °C
Standards: The product does not use any rawmaterials of animal or biological origin and therefore does not have TSE/BSE.
The polymer raw materials used for coating plates were positively evaluated for quality consistency using GPC, GC-MS and NMR spectroscopy.
The sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free multi-well plates were coated using pipetting robots. Coated plates were sterilised by electron beam irradiation.
Verpackung: Bag of 4 plates per pack.
Bitte beachten Research use only. Not intended for diagnostic or therapeutic use.