Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse) NF, ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Kaliumphosphat monobasisch

7390-19EA 0 EUR
7390-19
Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse) NF, ACS, J.T.Baker®
Kaliumdihydrogenphosphat
Formel: KH₂PO₄
Molecular Weight: 136.09 g/mol
Siedepunkt: 100
Schmelzpunkt: 253
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 98.0 - 100.5 %
NF - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
NF - Arsenic (As) ≤3 ppm
NF - Fluoride (F) ≤10 mg/kg
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Insoluble Substances ≤0.2 %
NF - Lead (Pb) ≤5 ppm
NF - Loss on Drying at 105°C ≤1.0 %
ACS - Assay (KH₂PO₄) (by acidimetry) ≥99.0 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Loss on Drying at 105°C ≤0.2 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - pH of 5% Solution at 25°C 4.1 - 4.5
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.003 %
Solubility (10 in 100) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR