Supelco® TraceCERT® Organic Reference Standard, Triflumuron
Lieferant: Merck
74022-50MG
74022-50MGEA 179 EUR
74022-50MG
Supelco® TraceCERT® Organic Reference Standard, Triflumuron
Standards Organische Standards
  • GHS signal: GHS06
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P260 - P271 - P284 - P304 + P340 + P310 - P403 + P233 - P405